Learn More
1,4-Benzoquinone, 98+%
1, 4-Benzoquinone, 99%, C6H4O2, CAS Number-106-51-4, 1,4-benzoquine, 1,4-benzoquinone, 2,5-cyclohexadiene-1,4-dione, benzoquinone, chinone, cyclohexadienedione, para-quinone, p-benzoquinone, p-quinone, quinone, 1kg, 98.5% min. (Iodometry), 07,609, 100.0, CHEBI:16509, Yellow to Green, 1.3100g/mL | CAS: 106-51-4 | C6H4O2 | 108.1 g/mol
Supplier: Thermo Scientific Chemicals 105630010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1, 4-Benzoquinone | |
| 98.5 | |
| 112°C to 116°C | |
| 1.3100g/mL | |
| 98.5% min. (Iodometry) | |
| C6H4O2 | |
| MFCD00001591 | |
| 07,609 | |
| 1.31 | |
| p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine | |
| AZQWKYJCGOJGHM-UHFFFAOYSA-N | |
| cyclohexa-2,5-diene-1,4-dione | |
| 4650 | |
| 108.1 | |
| Powder |
| 106-51-4 | |
| 100.0 | |
| Yellow to Green | |
| Authentic | |
| Plastic bottle | |
| C6H4(=O)2 | |
| 1 kg | |
| 01,49; 07,55 | |
| 15,8191 | |
| Solubility in water: 10g/L water (25°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether | |
| C1=CC(=O)C=CC1=O | |
| 108.1 | |
| CHEBI:16509 | |
| 99% |
Chemical Identifiers
| 106-51-4 | |
| 108.1 | |
| AZQWKYJCGOJGHM-UHFFFAOYSA-N | |
| 4650 | |
| cyclohexa-2,5-diene-1,4-dione |
| C6H4O2 | |
| MFCD00001591 | |
| p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine | |
| CHEBI:16509 | |
| C1=CC(=O)C=CC1=O |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life.
Toxic if swallowed.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-405-2
RUO – Research Use Only