missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzoquinone, 98+%

Catalog No. AC105635000
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105635000 500 g Plastic bottle
AC105631000 100 g Plastic bottle
AC105630010 1 kg Plastic bottle
AC105630051 5 kg Plastic drum
AC105630250 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC105635000 Supplier Thermo Scientific Chemicals Supplier No. 105635000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

1, 4-Benzoquinone, 99%, C6H4O2, CAS Number-106-51-4, 1,4-benzoquine, 1,4-benzoquinone, 2,5-cyclohexadiene-1,4-dione, benzoquinone, chinone, cyclohexadienedione, para-quinone, p-benzoquinone, p-quinone, quinone, 1kg, 98.5% min. (Iodometry), 07,609, 100.0, CHEBI:16509, Yellow to Green, 1.3100g/mL | CAS: 106-51-4 | C6H4O2 | 108.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-51-4
Molecular Formula C6H4O2
Molecular Weight (g/mol) 108.1
MDL Number MFCD00001591
InChI Key AZQWKYJCGOJGHM-UHFFFAOYSA-N
Synonym p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine
PubChem CID 4650
ChEBI CHEBI:16509
IUPAC Name cyclohexa-2,5-diene-1,4-dione
SMILES C1=CC(=O)C=CC1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 112°C to 116°C
Color Yellow to Green
Density 1.3100g/mL
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodometry)
Packaging Plastic bottle
Linear Formula C6H4(=O)2
Quantity 500 g
Beilstein 07,609
Fieser 01,49; 07,55
Specific Gravity 1.31
Merck Index 15,8191
Solubility Information Solubility in water: 10g/L water (25°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether
Formula Weight 108.1
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1, 4-Benzoquinone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-405-2

RUO – Research Use Only