missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxane-6-carboxylic Acid 98.0+%, TCI America™

Catalog No. B37645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37645G 5 g
B376425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37645G Supplier TCI America Supplier No. B37645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4442-54-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00463509
InChI Key JWZQJTGQFHIRFQ-UHFFFAOYSA-N
Synonym 1,4-benzodioxane-6-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-6-carboxylic acid, 3,4-ethylenedioxy benzoic acid, 2,3-dihydro-benzo 1,4 dioxine-6-carboxylic acid, 1,4-benzodioxin-6-carboxylic acid, 2,3-dihydro, 3,4-ethylenedioxybenzoic acid, 1,4-benzodioxan-6-carboxylic acid, 2,3-dihydro-1,4-benzodioxin-6-carboxylic acid, 2,3-dihydro-1,4-benzodioxine-6-carboxylicacid, 6-carboxy-2,3-dihydro-1,4-benzodioxine
PubChem CID 2758833
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6-carboxylic acid
SMILES C1COC2=C(O1)C=CC(=C2)C(=O)O

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Benzodioxane-6-carboxylic Acid