Learn More
1,4-Benzodioxane-6-carboxaldehyde, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1869606
Description
1,4-Benzodioxan-6-carboxaldehyde finds its uses as building block in the synthesis of tetrahydroisoquinolinones, in the preparation of benzofuran analog, a potential inhibitor of CAMP-specific phosphodiesterase type IV, in the synthesis of (5Z)-5-(2,3-dihydro-1,4-benzodioxan-6-ylmethylene)-1-methyl-2-thioxoimidazolidin-4-one. It was screened as a fragment for inhibitors of PDEA using enthalpy arrays and X-ray crystallography. 1,4-Benzodioxane-6-carboxaldehyde have also been used as an intermediate in the synthesis of a new class of Dopamine D3 and D4 receptor antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Benzodioxan-6-carboxaldehyde finds its uses as building block in the synthesis of tetrahydroisoquinolinones, in the preparation of benzofuran analog, a potential inhibitor of CAMP-specific phosphodiesterase type IV, in the synthesis of (5Z)-5-(2,3-dihydro-1,4-benzodioxan-6-ylmethylene)-1-methyl-2-thioxoimidazolidin-4-one. It was screened as a fragment for inhibitors of PDEA using enthalpy arrays and X-ray crystallography. 1,4-Benzodioxane-6-carboxaldehyde have also been used as an intermediate in the synthesis of a new class of Dopamine D3 and D4 receptor antagonists.
Solubility
Solubility in methanol is almost transparent.
Notes
Store away from air, oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 1,4-Benzodioxane-6-carboxaldehyde | |
| 48°C to 52°C | |
| >110°C (230°F) | |
| MFCD00010092 | |
| Air Sensitive | |
| CWKXDPPQCVWXAG-UHFFFAOYSA-N | |
| 2,3-dihydro-1,4-benzodioxine-6-carbaldehyde | |
| 248127 | |
| 99% |
| 29668-44-8 | |
| 104°C to 106°C (15 mmHg) | |
| C9H8O3 | |
| 5 g | |
| 1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde | |
| C1COC2=C(O1)C=CC(=C2)C=O | |
| 164.16 | |
| 164.16 |
Chemical Identifiers
| 29668-44-8 | |
| 164.16 | |
| CWKXDPPQCVWXAG-UHFFFAOYSA-N | |
| 248127 | |
| C1COC2=C(O1)C=CC(=C2)C=O |
| C9H8O3 | |
| MFCD00010092 | |
| 1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde | |
| 2,3-dihydro-1,4-benzodioxine-6-carbaldehyde |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 249-763-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only