missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-6-carboxaldehyde, 98%, Thermo Scientific™

Catalog No. p-3756447
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398210010 1g Glass bottle
AC398210050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398210010 Supplier Thermo Scientific Chemicals Supplier No. 398210010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29668-44-8
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00010092
InChI Key CWKXDPPQCVWXAG-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde
PubChem CID 248127
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
SMILES C1COC2=C(O1)C=CC(=C2)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Boiling Point 105°C (20.0 hPa)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1g
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material 1, 4-Benzodioxan-6-carboxaldehyde
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 249-763-3

RUO – Research Use Only