missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-6-carboxaldehyde, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 398210050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC398210050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

29668-44-8
164.16
CWKXDPPQCVWXAG-UHFFFAOYSA-N
248127
C1COC2=C(O1)C=CC(=C2)C=O
C9H8O3
MFCD00010092
1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde
2,3-dihydro-1,4-benzodioxine-6-carbaldehyde

Specifications

1, 4-Benzodioxan-6-carboxaldehyde
97.5
105°C (20.0 hPa)
Authentic
Glass bottle
5g
1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde
C1COC2=C(O1)C=CC(=C2)C=O
164.16
164.16
29668-44-8
100.0
110°C
97.5% min. (GC)
C9H8O3
MFCD00010092
CWKXDPPQCVWXAG-UHFFFAOYSA-N
2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
248127
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 249-763-3

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...