missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-6-amine, 99%

Catalog No. p-7043583
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0142106 5 g
AAL0142114 25 g
AAL0142122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0142106 Supplier Thermo Scientific Chemicals Supplier No. L0142106
Only null left

CAS: 22013-33-8 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22013-33-8
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00006824
InChI Key BZKOZYWGZKRTIB-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-6-amine, 3,4-ethylenedioxyaniline, 6-amino-1,4-benzodioxane, 2,3-dihydrobenzo b 1,4 dioxin-6-amine, 6-amino-1,4-benzodioxan, 1,4-benzodioxan-6-ylamine, 1,4-benzodioxin-6-amine, 2,3-dihydro, 3,4-ethylenedioxy aniline, 2,3-dihydro-benzo 1,4 dioxin-6-ylamine, 2,3-dihydro-1,4-benzodioxin-6-amin
PubChem CID 89148
IUPAC Name 2,3-dihydro-1,4-benzodioxin-6-amine
SMILES NC1=CC=C2OCCOC2=C1

Specifications

Melting Point 26°C to 30°C
Density 1.231
Boiling Point 116°C to 118°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.599
Quantity 5 g
UN Number UN2811
Beilstein 6447
Sensitivity Air and light sensitive
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 1,4-Benzodioxan-6-amine
Show More Show Less
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 244-718-4
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only