missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenedimethanethiol 98.0+%, TCI America™

Catalog No. p-7119091
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15211G 1 g
B15215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15211G Supplier TCI America Supplier No. B15211G
Only null left

Chemical Identifiers

CAS 105-09-9
Molecular Formula C8H10S2
Molecular Weight (g/mol) 170.29
MDL Number MFCD00004872
InChI Key IYPNRTQAOXLCQW-UHFFFAOYSA-N
PubChem CID 66043
IUPAC Name [4-(sulfanylmethyl)phenyl]methanethiol
SMILES SCC1=CC=C(CS)C=C1

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 156°C
Quantity 1 g
Synonym alpha,alpha′C-Dimercapto-p-xylene, 1,4-Bis(mercaptomethyl)benzene
Formula Weight 170.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Benzenedimethanethiol