missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenediboronic acid bis(pinacol) ester, 97%

Catalog No. p-7034059
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5170703 1 g
AAH5170706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5170703 Supplier Thermo Scientific Chemicals Supplier No. H5170703
Only null left

CAS: 99770-93-1 | C18H28B2O4 | 330.038 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99770-93-1
Molecular Formula C18H28B2O4
Molecular Weight (g/mol) 330.038
MDL Number MFCD08276852
InChI Key UOJCDDLTVQJPGH-UHFFFAOYSA-N
Synonym 1,4-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, 1,4-benzenediboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3,2-dioxaborolane, 1,4-phenylenediboronic acid, pinacol ester, 2,2'-1,4-phenylene bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2,2'-1,4-phenylene bis 4,4,5,5-tetramethyl, 4,4,5,5-tetramethyl-2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3,2-dioxaborolane, 1,4-benzenediboronic acid dipinacol ester, amtb185, 1,4-phenylenediboronic acid pinacol ester
PubChem CID 10592495
IUPAC Name 4,4,5,5-tetramethyl-2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)B3OC(C(O3)(C)C)(C)C

Specifications

Quantity 1 g
Formula Weight 330.03
Percent Purity 97%
Chemical Name or Material 1,4-Benzenediboronic acid bis(pinacol) ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only