missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenediboronic acid, 96%

Catalog No. AAB2406414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2406414 25 g
AAB2406403 1 g
AAB2406406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2406414 Supplier Thermo Scientific Chemicals Supplier No. B2406414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4612-26-4 | C6H8B2O4 | 165.746 g/mol

It is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.

Solubility
Soluble in water 2.5%.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 4612-26-4
Molecular Formula C6H8B2O4
Molecular Weight (g/mol) 165.746
MDL Number MFCD00236018
InChI Key BODYVHJTUHHINQ-UHFFFAOYSA-N
Synonym 1,4-benzenediboronic acid, 1,4-phenylenediboronic acid, 1,4-phenylenebisboronic acid, benzene-1,4-diboronic acid, 4-boronophenyl boronic acid, p-benzenediboronic acid, p-phenylenediboronic acid, 1,4-phenylenebis boronic acid, 1,4 phenyl diboronic acid, boronic acid, 1,4-phenylenebis
PubChem CID 230478
IUPAC Name (4-boronophenyl)boronic acid
SMILES B(C1=CC=C(C=C1)B(O)O)(O)O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 2836921
Solubility Information Soluble in water 2.5%.
Formula Weight 165.75
Percent Purity 96%
Chemical Name or Material 1,4-Benzenediboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only