missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Anthraquinone 97.0+%, TCI America™
$154.71 - $631.92
Chemical Identifiers
| CAS | 635-12-1 |
|---|---|
| Molecular Formula | C14H8O2 |
| Molecular Weight (g/mol) | 208.216 |
| MDL Number | MFCD00013724 |
| InChI Key | LSOTZYUVGZKSHR-UHFFFAOYSA-N |
| Synonym | 1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione |
| PubChem CID | 69457 |
| IUPAC Name | anthracene-1,4-dione |
| SMILES | C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1 |
Chemical Identifiers
| 635-12-1 | |
| 208.216 | |
| LSOTZYUVGZKSHR-UHFFFAOYSA-N | |
| 69457 | |
| C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1 |
| C14H8O2 | |
| MFCD00013724 | |
| 1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione | |
| anthracene-1,4-dione |
Specifications
| 635-12-1 | |
| C14H8O2 | |
| 1 g | |
| LSOTZYUVGZKSHR-UHFFFAOYSA-N | |
| anthracene-1,4-dione | |
| 69457 | |
| ≥95.0% (GC) | |
| 1,4-Anthraquinone |
| 218°C | |
| MFCD00013724 | |
| 1,4-anthraquinone, 1,4-anthracenedione, 1,4-dioxoanthracene, 1,4-anthracenequinone, acmc-1b3md, 1,4-dihydroanthracene-1,4-dione | |
| C1=CC=C2C=C3C(=O)C=CC(=O)C3=CC2=C1 | |
| 208.216 | |
| 208.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No