missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Amino-2-hydroxyphenyl)ethan-1-one, Tech., Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 151.16 |
| Percent Purity | Tech. |
| Concentration or Composition (by Analyte or Components) | Technical Grade |
| Chemical Name or Material | 1-(4-Amino-2-hydroxyphenyl)ethan-1-one |
Safety and Handling
| Hazard Category | Warning |