missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Acetylpiperidino)ethan-1-one, 97%, Thermo Scientific™

Catalog No. p-4391146
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CD11879DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CD11879DA Supplier Thermo Scientific Chemicals Supplier No. CD11879DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 162368-01-6
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.22
MDL Number MFCD00106774
InChI Key VFZXZEJGLKOTBW-UHFFFAOYSA-N
Synonym 1-4-acetylpiperidino ethan-1-one, 1-1-acetylpiperidin-4-yl ethanone, 1,4-diacetylpiperidine, 1-4-acetylpiperidin-1-yl ethan-1-one, 1,1'-piperidine-1,4-diyl diethanone, 1-1-acetyl-piperidin-4-yl-ethanone, piperidine, 1,4-diacetyl-9ci, 1-4-acetylpiperidin-1-yl ethanone, ethanone,1-1-acetyl-4-piperidinyl, piperidine, 1,4-diacetyl
PubChem CID 2777250
IUPAC Name 1-(1-acetylpiperidin-4-yl)ethanone
SMILES CC(=O)C1CCN(CC1)C(C)=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 169.22
Percent Purity 97%
Chemical Name or Material 1-(4-Acetylpiperidino)ethan-1-one
Hazard Category Warning