missing translation for 'onlineSavingsMsg'
Learn More

1,4,8,11-Tetraazacyclotetradecane, 98+%

Catalog No. p-7081514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
AAA1151603 1 g
AAA1151606 5 g
AAA1151614 25 g
3 options

Catalog No. AAA1151603

Supplier: Thermo Scientific Chemicals A1151603

Only null left

CAS: 295-37-4 | C10H24N4 | 200.33 g/mol

1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 295-37-4
Molecular Formula C10H24N4
Molecular Weight (g/mol) 200.33
MDL Number MFCD00005105
InChI Key MDAXKAUIABOHTD-UHFFFAOYSA-N
Synonym 1,4,8,11-tetraazacyclotetradecane, cyclam, 14 anen4, 1,4,8,11-tetraazacyclo-tetradecane, 1,4,8,11-tetraazacyclotetradecane cyclam, ips2, acmc-209h9n, 5-26-11-00027 beilstein handbook reference, 1,8,11-tetraazacyclotetradecane, 1,4,8,11tetraaza-cyclotetradecane
PubChem CID 64964
ChEBI CHEBI:37401
IUPAC Name 1,4,8,11-tetrazacyclotetradecane
SMILES C1CNCCNCCCNCCNC1

Specifications

Melting Point 184°C to 188°C
Quantity 1 g
Beilstein 111811
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water.
Formula Weight 200.33
Percent Purity ≥98%
Chemical Name or Material 1,4,8,11-Tetraazacyclotetradecane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-039-1
RTECSNumber XA5254800
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only