missing translation for 'onlineSavingsMsg'
Learn More

1,4,8,11-Tetraazacyclotetradecane 98.0+%, TCI America™

Catalog No. p-7136863
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15971G 1 g
T15975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15971G Supplier TCI America Supplier No. T15971G
Only null left

Chemical Identifiers

CAS 295-37-4
Molecular Formula C10H24N4
Molecular Weight (g/mol) 200.33
MDL Number MFCD00005105
InChI Key MDAXKAUIABOHTD-UHFFFAOYSA-N
Synonym 1,4,8,11-tetraazacyclotetradecane, cyclam, 14 anen4, 1,4,8,11-tetraazacyclo-tetradecane, 1,4,8,11-tetraazacyclotetradecane cyclam, ips2, acmc-209h9n, 5-26-11-00027 beilstein handbook reference, 1,8,11-tetraazacyclotetradecane, 1,4,8,11tetraaza-cyclotetradecane
PubChem CID 64964
ChEBI CHEBI:37401
IUPAC Name 1,4,8,11-tetrazacyclotetradecane
SMILES C1CNCCNCCCNCCNC1

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 1 g
Formula Weight 200.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,8,11-Tetraazacyclotetradecane
RTECSNumber XA5254800
TSCA No