missing translation for 'onlineSavingsMsg'
Learn More

1,4,7-Trimethyldiethylenetriamine, 95%

Catalog No. AAH66905MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66905MD 250 mg
AAH6690503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66905MD Supplier Thermo Scientific Chemicals Supplier No. H66905MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 105-84-0 | C7H19N3 | 145.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-84-0
Molecular Formula C7H19N3
Molecular Weight (g/mol) 145.25
MDL Number MFCD00025637
InChI Key ODZZIKZQNODXFS-UHFFFAOYSA-N
Synonym 1,4,7-trimethyldiethylenetriamine, n1,n2-dimethyl-n1-2-methylamino ethyl ethane-1,2-diamine, n,n',n-trimethyldiethylenetriamine, 1,2-ethanediamine, n,n'-dimethyl-n-2-methylamino ethyl, n,n'-dimethyl-n-2-methylamino ethyl ethylenediamine, methyl 2-methyl 2-methylamino ethyl amino ethyl amine, 1,2-ethanediamine, n1,n2-dimethyl-n1-2-methylamino ethyl
PubChem CID 66930
IUPAC Name N,N'-dimethyl-N'-[2-(methylamino)ethyl]ethane-1,2-diamine
SMILES CNCCN(C)CCNC

Specifications

Color Yellow
Linear Formula (CH3NHCH2CH2)2NCH3
Quantity 250 mg
Beilstein 1736861
Formula Weight 145.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1,4,7-Trimethyldiethylenetriamine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 203-337-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only