missing translation for 'onlineSavingsMsg'
Learn More

1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane

CAS: 157599-02-5 | C16H32N8O4 | 400.48 g/mol

Supplier:  Thermo Scientific Chemicals H2679203

 View more versions of this product

Catalog No. AAH2679203


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane
C16H32N8O4
1 g
2,2',2,2'-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetrayl tetraacetamide, dotam, 1,4,7,10-tetrakis aminocarbonylmethyl-1,4,7,10-tetraazacyclododecane, 2-4,7,10-tris carbamoylmethyl-1,4,7,10-tetraazacyclododecan-1-yl acetamide, acmc-209dgc, cyclen-n,n',n',n-tetraacetamide, 1,4,7,10-tetraazacyclododecane 1,4,7,10-tetraacetamide, 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetamide, 1,4,7,10-tetrakis carbamoylmethyl-1,4,7,10-tetraazacyclododecane
NC(=O)CN1CCN(CC(N)=O)CCN(CC(N)=O)CCN(CC(N)=O)CC1
400.48
400.48
157599-02-5
MFCD09263318
Hygroscopic
FQIHLPGWBOBPSG-UHFFFAOYSA-N
2-[4,7,10-tris(2-amino-2-oxoethyl)-1,4,7,10-tetrazacyclododec-1-yl]acetamide
11560395

Chemical Identifiers

157599-02-5
400.48
FQIHLPGWBOBPSG-UHFFFAOYSA-N
11560395
NC(=O)CN1CCN(CC(N)=O)CCN(CC(N)=O)CCN(CC(N)=O)CC1
C16H32N8O4
MFCD09263318
2,2',2,2'-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetrayl tetraacetamide, dotam, 1,4,7,10-tetrakis aminocarbonylmethyl-1,4,7,10-tetraazacyclododecane, 2-4,7,10-tris carbamoylmethyl-1,4,7,10-tetraazacyclododecan-1-yl acetamide, acmc-209dgc, cyclen-n,n',n',n-tetraacetamide, 1,4,7,10-tetraazacyclododecane 1,4,7,10-tetraacetamide, 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetamide, 1,4,7,10-tetrakis carbamoylmethyl-1,4,7,10-tetraazacyclododecane
2-[4,7,10-tris(2-amino-2-oxoethyl)-1,4,7,10-tetrazacyclododec-1-yl]acetamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only