missing translation for 'onlineSavingsMsg'
Learn More

1,4,7,10-Tetraazacyclotridecane

CAS: 295-14-7 | C9H22N4 | 186.303 g/mol

$129.22 - $349.53

Chemical Identifiers

CAS 295-14-7
Molecular Formula C9H22N4
Molecular Weight (g/mol) 186.303
MDL Number MFCD06410986
InChI Key LADZJJOUGVGJHM-UHFFFAOYSA-N
Synonym 1,4,7,10-tetraazacyclotridecane, homocyclen, acmc-20ap3n, 13 anen4
PubChem CID 11478723
IUPAC Name 1,4,7,10-tetrazacyclotridecane
SMILES C1CNCCNCCNCCNC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH26671MD
View Documents
Thermo Scientific Chemicals
H26671MD
250 mg
Each for $129.22
Only null left
 
AAH2667103
View Documents
Thermo Scientific Chemicals
H2667103
1 g
Each for $349.53
Only null left
 
Description

Description

1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

295-14-7
186.303
LADZJJOUGVGJHM-UHFFFAOYSA-N
11478723
C1CNCCNCCNCCNC1
C9H22N4
MFCD06410986
1,4,7,10-tetraazacyclotridecane, homocyclen, acmc-20ap3n, 13 anen4
1,4,7,10-tetrazacyclotridecane

Specifications

295-14-7
MFCD06410986
Hygroscopic
Soluble in water.
C1CNCCNCCNCCNC1
186.303
186.3
C9H22N4
250 mg
1,4,7,10-tetraazacyclotridecane, homocyclen, acmc-20ap3n, 13 anen4
LADZJJOUGVGJHM-UHFFFAOYSA-N
1,4,7,10-tetrazacyclotridecane
11478723
1,4,7,10-Tetraazacyclotridecane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only