missing translation for 'onlineSavingsMsg'
Learn More

1,4,7,10-Tetraazacyclotridecane

Catalog No. AAH26671MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26671MD 250 mg
AAH2667103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26671MD Supplier Thermo Scientific Chemicals Supplier No. H26671MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 295-14-7 | C9H22N4 | 186.303 g/mol

1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 295-14-7
Molecular Formula C9H22N4
Molecular Weight (g/mol) 186.303
MDL Number MFCD06410986
InChI Key LADZJJOUGVGJHM-UHFFFAOYSA-N
Synonym 1,4,7,10-tetraazacyclotridecane, homocyclen, acmc-20ap3n, 13 anen4
PubChem CID 11478723
IUPAC Name 1,4,7,10-tetrazacyclotridecane
SMILES C1CNCCNCCNCCNC1

Specifications

Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 186.3
Chemical Name or Material 1,4,7,10-Tetraazacyclotridecane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only