missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4,7,10-Tetraazacyclododecane 97.0+%, TCI America™
$52.19 - $154.71
Chemical Identifiers
| CAS | 294-90-6 |
|---|---|
| Molecular Formula | C8H20N4 |
| Molecular Weight (g/mol) | 172.276 |
| InChI Key | QBPPRVHXOZRESW-UHFFFAOYSA-N |
| Synonym | cyclen, 1,4,7,10-tetraazacyclododecane, 12 anen4, 1,4,7,10tetraaza-cyclododecane, 5-26-11-00023 beilstein handbook reference, 1,4,7,10-tetracyclododecane, 1,4,7,10-tetraazacyclododecane cyclen, tetraaza-12-crown-4, pubchem23801 |
| PubChem CID | 64963 |
| ChEBI | CHEBI:37391 |
| IUPAC Name | 1,4,7,10-tetrazacyclododecane |
| SMILES | C1CNCCNCCNCCN1 |
Chemical Identifiers
| 294-90-6 | |
| 172.276 | |
| cyclen, 1,4,7,10-tetraazacyclododecane, 12 anen4, 1,4,7,10tetraaza-cyclododecane, 5-26-11-00023 beilstein handbook reference, 1,4,7,10-tetracyclododecane, 1,4,7,10-tetraazacyclododecane cyclen, tetraaza-12-crown-4, pubchem23801 | |
| CHEBI:37391 | |
| C1CNCCNCCNCCN1 |
| C8H20N4 | |
| QBPPRVHXOZRESW-UHFFFAOYSA-N | |
| 64963 | |
| 1,4,7,10-tetrazacyclododecane |
Specifications
| 294-90-6 | |
| White-Yellow | |
| 1 g | |
| cyclen, 1,4,7,10-tetraazacyclododecane, 12 anen4, 1,4,7,10tetraaza-cyclododecane, 5-26-11-00023 beilstein handbook reference, 1,4,7,10-tetracyclododecane, 1,4,7,10-tetraazacyclododecane cyclen, tetraaza-12-crown-4, pubchem23801 | |
| C1CNCCNCCNCCN1 | |
| 172.276 | |
| CHEBI:37391 | |
| ≥97.0% (GC,T) | |
| 1,4,7,10-Tetraazacyclododecane |
| 111°C | |
| C8H20N4 | |
| 3259 | |
| QBPPRVHXOZRESW-UHFFFAOYSA-N | |
| 1,4,7,10-tetrazacyclododecane | |
| 64963 | |
| 172.28 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : XA5253000
TSCA : No