missing translation for 'onlineSavingsMsg'
Learn More

1,4,7,10-Tetraazacyclododecane 97.0+%, TCI America™

Catalog No. T18745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18745G 5 g
T18741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18745G Supplier TCI America Supplier No. T18745G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 294-90-6
Molecular Formula C8H20N4
Molecular Weight (g/mol) 172.276
InChI Key QBPPRVHXOZRESW-UHFFFAOYSA-N
Synonym cyclen, 1,4,7,10-tetraazacyclododecane, 12 anen4, 1,4,7,10tetraaza-cyclododecane, 5-26-11-00023 beilstein handbook reference, 1,4,7,10-tetracyclododecane, 1,4,7,10-tetraazacyclododecane cyclen, tetraaza-12-crown-4, pubchem23801
PubChem CID 64963
ChEBI CHEBI:37391
IUPAC Name 1,4,7,10-tetrazacyclododecane
SMILES C1CNCCNCCNCCN1

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 5 g
UN Number 3259
Formula Weight 172.28
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,7,10-Tetraazacyclododecane
RTECSNumber XA5253000
TSCA No