missing translation for 'onlineSavingsMsg'
Learn More

1,4,5,8-Naphthalenetetracarboxdiimide 95.0+%, TCI America™

Catalog No. N05365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05365G 5 g
N05361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05365G Supplier TCI America Supplier No. N05365G
Only null left

Chemical Identifiers

CAS 5690-24-4
Molecular Formula C14H6N2O4
Molecular Weight (g/mol) 266.21
MDL Number MFCD00047649
InChI Key BODUWJSFPLUDMP-UHFFFAOYSA-N
PubChem CID 79771
IUPAC Name 6,13-diazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
SMILES O=C1NC(=O)C2=CC=C3C(=O)NC(=O)C4=CC=C1C2=C34

Specifications

Quantity 5 g
Formula Weight 266.21
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,5,8-Naphthalenetetracarboxdiimide