missing translation for 'onlineSavingsMsg'
Learn More

1,4,5,6-Tetrahydro-6-oxopyridazine-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. T29551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29551G 1 g
T29555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29551G Supplier TCI America Supplier No. T29551G

Chemical Identifiers

CAS 27372-38-9
Molecular Formula C5H6N2O3
Molecular Weight (g/mol) 142.114
MDL Number MFCD00052030
InChI Key VUADWGRLHPTYPI-UHFFFAOYSA-N
Synonym 6-oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid, 1,4,5,6-tetrahydro-6-oxopyridazine-3-carboxylic acid, unii-ot09ztp07e, ot09ztp07e, 6-oxo-1,4,5,6-tetrahydro-pyridazine-3-carboxylic acid, 3-pyridazinecarboxylic acid, 1,4,5,6-tetrahydro-6-oxo, 1,4,5,6-tetrahydro-6-oxo-3-pyridazinecarboxylic acid, 6-oxo-1,4,5,6-tetrahydropyridazin-3-carboxylic acid, 6-oxo-1,4,5-trihydropyridazine-3-carboxylic acid, pubchem19274
PubChem CID 99621
IUPAC Name 6-oxo-4,5-dihydro-1H-pyridazine-3-carboxylic acid
SMILES C1CC(=O)NN=C1C(=O)O

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 1 g
Formula Weight 142.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,5,6-Tetrahydro-6-oxopyridazine-3-carboxylic Acid