missing translation for 'onlineSavingsMsg'
Learn More

1,4,5,6-Tetrahydro-5,6-dioxo-2,3-pyrazinedicarbonitrile 98.0+%, TCI America™

Catalog No. T147210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T147210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T147210G Supplier TCI America Supplier No. T147210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36023-64-0
Molecular Formula C6H2N4O2
Molecular Weight (g/mol) 162.108
MDL Number MFCD00191690
InChI Key OTVDGBINMCDFTF-UHFFFAOYSA-N
Synonym 2,3-Dicyano-1,4,5,6-tetrahydropyrazine-5,6-dione
PubChem CID 595471
IUPAC Name 5,6-dioxo-1,4-dihydropyrazine-2,3-dicarbonitrile
SMILES C(#N)C1=C(NC(=O)C(=O)N1)C#N

Specifications

Color Yellow
Quantity 10 g
Formula Weight 162.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,5,6-Tetrahydro-5,6-dioxo-2,3-pyrazinedicarbonitrile