missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(4-Morpholinyl)phenyl]guanidine, 98%

Catalog No. AAH3372603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3372603 1 g
AAH3372606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3372603 Supplier Thermo Scientific Chemicals Supplier No. H3372603

CAS: 247234-41-9 | C11H16N4O | 220.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 247234-41-9
Molecular Formula C11H16N4O
Molecular Weight (g/mol) 220.28
MDL Number MFCD11986905
InChI Key ZVZJREQBRCRGLM-UHFFFAOYSA-N
Synonym 1-4-morpholinophenyl guanidine, n-4-morpholin-4-ylphenyl guanidine, 4-morpholinophenylguanidine, 4-morpholino phenylguanidine, n-4-morpholin-4-yl-phenyl-guanidine, n-4-morpholin-4-yl phenyl guanidine, 1-4-4-morpholinyl phenyl guanidine, 1-4-morpholin-4-yl phenyl guanidine, 2-4-morpholin-4-yl phenyl guanidine
PubChem CID 10176830
IUPAC Name 2-(4-morpholin-4-ylphenyl)guanidine
SMILES NC(N)=NC1=CC=C(C=C1)N1CCOCC1

Specifications

Melting Point ∼220°C (decomposition)
Density 1.309
Quantity 1 g
Formula Weight 220.27
Percent Purity 98%
Chemical Name or Material 1-[4-(4-Morpholinyl)phenyl]guanidine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only