missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC33939DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC33939DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

852227-96-4
287.21
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 4-1-piperidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-piperidine, piperidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl, amtb222, 4-piperidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl piperidine
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
C17H26BNO2
OTOKWHGMHAAFRM-UHFFFAOYSA-N
4961250
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCCCC3

Specifications

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
97%
C17H26BNO2
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 4-1-piperidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-piperidine, piperidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl, amtb222, 4-piperidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl piperidine
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCCCC3
287.21
287.21
852227-96-4
Amber Glass Bottle
5g
OTOKWHGMHAAFRM-UHFFFAOYSA-N
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
4961250
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents