missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(2-Phenylethyl)benzyl]naphthalene 98.0+%, TCI America™

Catalog No. P118125G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P118125G 25 g
P11811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P118125G Supplier TCI America Supplier No. P118125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127833-53-8
Molecular Formula C25H22
Molecular Weight (g/mol) 322.451
MDL Number MFCD00191572
InChI Key UKRRCRMJNMWWMZ-UHFFFAOYSA-N
Synonym 1-[4-(2-Phenethyl)benzyl]naphthalene
PubChem CID 14833659
IUPAC Name 1-[[4-(2-phenylethyl)phenyl]methyl]naphthalene
SMILES C1=CC=C(C=C1)CCC2=CC=C(C=C2)CC3=CC=CC4=CC=CC=C43

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 25 g
Formula Weight 322.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-[4-(2-Phenylethyl)benzyl]naphthalene