missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(Triethoxysilyl)propyl]urea (40-52% in Methanol), TCI America™

Catalog No. U004825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U004825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U004825ML Supplier TCI America Supplier No. U004825ML
Only null left

Chemical Identifiers

CAS 23779-32-0
Molecular Formula C10H24N2O4Si
Molecular Weight (g/mol) 264.397
MDL Number MFCD00047877
InChI Key LVNLBBGBASVLLI-UHFFFAOYSA-N
Synonym Triethoxy-3-ureidopropylsilane, 3-Ureidopropyltriethoxysilane
PubChem CID 90258
IUPAC Name 3-triethoxysilylpropylurea
SMILES CCO[Si](CCCNC(=O)N)(OCC)OCC

Specifications

Color Colorless
Quantity 25 mL
UN Number 1993
Formula Weight 264.40
Physical Form Liquid
Chemical Name or Material 1-[3-(Triethoxysilyl)propyl]urea (40-52% in Methanol)
EINECSNumber (2)-2968
TSCA Yes