missing translation for 'onlineSavingsMsg'
Learn More

1,3-Thiazol-2-ylmethanol, 97+%, Thermo Scientific™

Catalog No. CC09509DE
Change view
Click to view available options
Quantity:
5g
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC09509DE 5g Amber Glass Bottle
CC09509EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC09509DE Supplier Thermo Scientific Chemicals Supplier No. CC09509DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14542-12-2
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
MDL Number MFCD06200855
InChI Key JNHDLNXNYPLBMJ-UHFFFAOYSA-N
Synonym thiazol-2-ylmethanol, 2-thiazolemethanol, thiazol-2-yl-methanol, 2-hydroxymethylthiazole, 2-hydroxymethyl thiazole, 2-hydroxymehtylthiazole, 1,3-thiazol-2-yl methanol, thiazol-2-yl methanol, thiazole-2-methanol, 2-thiazolylmethanol
PubChem CID 2795213
IUPAC Name (1,3-thiazol-2-yl)methanol
SMILES OCC1=NC=CS1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5g
Formula Weight 115.16
Percent Purity 97%
Chemical Name or Material 1, 3-Thiazol-2-ylmethanol
Hazard Category Warning