missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3-Sulfopropyl)pyridinium Hydroxide Inner Salt 98.0+%, TCI America™
$35.41 - $167.76
Chemical Identifiers
| CAS | 15471-17-7 |
|---|---|
| Molecular Formula | C8H11NO3S |
| Molecular Weight (g/mol) | 201.24 |
| MDL Number | MFCD00064468 |
| InChI Key | REEBJQTUIJTGAL-UHFFFAOYSA-N |
| Synonym | 3-pyridin-1-ium-1-yl propane-1-sulfonate, ndsb-201, 1-3-sulphonatopropyl pyridinium, unii-qn4i6ai9ek, 3-1-pyridinio-1-propanesulfonate, 1-pyridiniumpropane-3-sulfonate, qn4i6ai9ek, 1-3-sulfopropyl pyridinium hydroxide, inner salt, pyridinium, 1-3-sulfopropyl-, inner salt, 1-3-sulfonatopropyl pyridin-1-ium |
| PubChem CID | 84929 |
| IUPAC Name | 1-(3-sulfonatopropyl)pyridin-1-ium |
| SMILES | [O-]S(=O)(=O)CCC[N+]1=CC=CC=C1 |
Chemical Identifiers
| 15471-17-7 | |
| 201.24 | |
| REEBJQTUIJTGAL-UHFFFAOYSA-N | |
| 84929 | |
| [O-]S(=O)(=O)CCC[N+]1=CC=CC=C1 |
| C8H11NO3S | |
| MFCD00064468 | |
| 3-pyridin-1-ium-1-yl propane-1-sulfonate, ndsb-201, 1-3-sulphonatopropyl pyridinium, unii-qn4i6ai9ek, 3-1-pyridinio-1-propanesulfonate, 1-pyridiniumpropane-3-sulfonate, qn4i6ai9ek, 1-3-sulfopropyl pyridinium hydroxide, inner salt, pyridinium, 1-3-sulfopropyl-, inner salt, 1-3-sulfonatopropyl pyridin-1-ium | |
| 1-(3-sulfonatopropyl)pyridin-1-ium |
Specifications
| 15471-17-7 | |
| White | |
| C8H11NO3S | |
| 25 g | |
| REEBJQTUIJTGAL-UHFFFAOYSA-N | |
| 1-(3-sulfonatopropyl)pyridin-1-ium | |
| 84929 | |
| ≥98.0% (HPLC,N) | |
| 1-(3-Sulfopropyl)pyridinium Hydroxide Inner Salt |
| 279°C | |
| 3 to 6 (10% H2O soln. 20°C) | |
| MFCD00064468 | |
| 3-pyridin-1-ium-1-yl propane-1-sulfonate, ndsb-201, 1-3-sulphonatopropyl pyridinium, unii-qn4i6ai9ek, 3-1-pyridinio-1-propanesulfonate, 1-pyridiniumpropane-3-sulfonate, qn4i6ai9ek, 1-3-sulfopropyl pyridinium hydroxide, inner salt, pyridinium, 1-3-sulfopropyl-, inner salt, 1-3-sulfonatopropyl pyridin-1-ium | |
| [O-]S(=O)(=O)CCC[N+]1=CC=CC=C1 | |
| 201.24 | |
| 201.24 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0758&(5)-3704
TSCA : Yes