missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Propylenediphosphonic Acid 98.0+%, TCI America™
$288.94 - $1017.78
Chemical Identifiers
| CAS | 4671-82-3 |
|---|---|
| Molecular Formula | C3H10O6P2 |
| Molecular Weight (g/mol) | 204.055 |
| InChI Key | DZQCAOQMXPROIJ-UHFFFAOYSA-N |
| Synonym | propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid |
| PubChem CID | 22023450 |
| IUPAC Name | 1-phosphonopropan-2-ylphosphonic acid |
| SMILES | CC(CP(=O)(O)O)P(=O)(O)O |
Chemical Identifiers
| 4671-82-3 | |
| 204.055 | |
| propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid | |
| 1-phosphonopropan-2-ylphosphonic acid |
| C3H10O6P2 | |
| DZQCAOQMXPROIJ-UHFFFAOYSA-N | |
| 22023450 | |
| CC(CP(=O)(O)O)P(=O)(O)O |
Specifications
| 4671-82-3 | |
| White | |
| 1 g | |
| propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid | |
| CC(CP(=O)(O)O)P(=O)(O)O | |
| 204.055 | |
| 204.05 | |
| Crystalline Powder |
| 174°C | |
| C3H10O6P2 | |
| 3261 | |
| DZQCAOQMXPROIJ-UHFFFAOYSA-N | |
| 1-phosphonopropan-2-ylphosphonic acid | |
| 22023450 | |
| ≥98.0% (T) | |
| 1,3-Propylenediphosphonic Acid |
Safety and Handling
TSCA : No