missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acide 1,3-propylène diphosphonique 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 174°C |
| Color | Blanc |
| Quantity | 5 g |
| UN Number | 3261 |
| Formula Weight | 204.05 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 1,3-Propylenediphosphonic Acid |
Sécurité et manipulation
| TSCA | No |