missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propylenediphosphonic Acid 98.0+%, TCI America™

Catalog No. P23041G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23041G 1 g
P23045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23041G Supplier TCI America Supplier No. P23041G

Chemical Identifiers

CAS 4671-82-3
Molecular Formula C3H10O6P2
Molecular Weight (g/mol) 204.055
InChI Key DZQCAOQMXPROIJ-UHFFFAOYSA-N
Synonym propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid
PubChem CID 22023450
IUPAC Name 1-phosphonopropan-2-ylphosphonic acid
SMILES CC(CP(=O)(O)O)P(=O)(O)O

Specifications

Melting Point 174°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 204.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Propylenediphosphonic Acid