missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanedithiol 97.0+%, TCI America™

Catalog No. P076325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P076325ML 25 mL
P0763100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P076325ML Supplier TCI America Supplier No. P076325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-80-8
Molecular Formula C3H8S2
Molecular Weight (g/mol) 108.217
MDL Number MFCD00004904
InChI Key ZJLMKPKYJBQJNH-UHFFFAOYSA-N
Synonym 1,3-propanedithiol, 1,3-dimercaptopropane, trimethylene dimercaptan, trimethylenedithiol, dithiotrimethyleneglycol, trimethylenedithioglycol, 1,3-propanedimercaptan, unii-r4luj82u52, fema no. 3588, 1,3-propane dithiol
PubChem CID 8013
ChEBI CHEBI:44864
IUPAC Name propane-1,3-dithiol
SMILES C(CS)CS

Specifications

Melting Point -79°C
Color Yellow
Boiling Point 170°C
Quantity 25 mL
Formula Weight 108.22
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,3-Propanedithiol
RTECSNumber TZ2585500
TSCA Yes