Learn More
1,3-Propanedithiol, 97%
CAS: 109-80-8 | C3H8S2 | 108.217 g/mol
Supplier: Thermo Scientific Chemicals A1526114
Description
1,3-Propanedithiol is used as a reagent in the preparation of thioketals and thioacetals. It acts as a flavoring agent. It is used as a precursor in the synthesis of cyclic dithioacetal (1,3-dithiane) derivatives of carbonyl compounds. It is involved in the preparation of diiron propanedithiolate hexacarbonyl by reacting reaction with triiron dodecacarbonyl. Further, it is used for the protection of aldehydes and ketones through their reversible formation of dithianes. In addition to this, it reacts with metal ions to form chelate rings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Propanedithiol is used as a reagent in the preparation of thioketals and thioacetals. It acts as a flavoring agent. It is used as a precursor in the synthesis of cyclic dithioacetal (1,3-dithiane) derivatives of carbonyl compounds. It is involved in the preparation of diiron propanedithiolate hexacarbonyl by reacting reaction with triiron dodecacarbonyl. Further, it is used for the protection of aldehydes and ketones through their reversible formation of dithianes. In addition to this, it reacts with metal ions to form chelate rings.
Solubility
Slightly soluble in water. Miscible with alcohol, ether, chloroform, etherMiscible with alcohol, ether, chloroform and ether. Slightly miscible with water.
Notes
Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
Specifications
| 1,3-Propanedithiol | |
| -79°C | |
| 168°C to 170°C | |
| Stench | |
| 1.54 | |
| 25 g | |
| 1071197 | |
| 1,3-propanedithiol, 1,3-dimercaptopropane, trimethylene dimercaptan, trimethylenedithiol, dithiotrimethyleneglycol, trimethylenedithioglycol, 1,3-propanedimercaptan, unii-r4luj82u52, fema no. 3588, 1,3-propane dithiol | |
| ZJLMKPKYJBQJNH-UHFFFAOYSA-N | |
| propane-1,3-dithiol | |
| 8013 | |
| 108.23 |
| 109-80-8 | |
| 1.077 | |
| 63°C (145°F) | |
| C3H8S2 | |
| MFCD00004904 | |
| UN3334 | |
| 14,7801 | |
| Slightly soluble in water; Miscible with alcohol,ether,chloroform,etherMiscible with alcohol,ether,chloroform and ether; Slightly miscible with water. | |
| C(CS)CS | |
| 108.217 | |
| CHEBI:44864 | |
| 97% |
Chemical Identifiers
| 109-80-8 | |
| 108.217 | |
| ZJLMKPKYJBQJNH-UHFFFAOYSA-N | |
| 8013 | |
| propane-1,3-dithiol |
| C3H8S2 | |
| MFCD00004904 | |
| 1,3-propanedithiol, 1,3-dimercaptopropane, trimethylene dimercaptan, trimethylenedithiol, dithiotrimethyleneglycol, trimethylenedithioglycol, 1,3-propanedimercaptan, unii-r4luj82u52, fema no. 3588, 1,3-propane dithiol | |
| CHEBI:44864 | |
| C(CS)CS |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 203-706-9
RTECSNumber : TZ2585500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only