missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Propanedisulfonic acid disodium salt, 99% (dry wt.)
Description
It is used as a reactant involved in synthesis of cadmium(II) bipyridine complex metal-organic frameworks with alkanedisulfonates, alkylenedisulfonate inorganic-organic hybrid materials and glycosaminoglycan mimetic involved in pharmacological studies for the treatment of amyloid A amyloidosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a reactant involved in synthesis of cadmium(II) bipyridine complex metal-organic frameworks with alkanedisulfonates, alkylenedisulfonate inorganic-organic hybrid materials and glycosaminoglycan mimetic involved in pharmacological studies for the treatment of amyloid A amyloidosis.
Solubility
Partially soluble in cold water, hot water.
Notes
Hygroscopic. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as a reactant involved in synthesis of cadmium(II) bipyridine complex metal-organic frameworks with alkanedisulfonates, alkylenedisulfonate inorganic-organic hybrid materials and glycosaminoglycan mimetic involved in pharmacological studies for the treatment of amyloid A amyloidosis.
Solubility
Partially soluble in cold water, hot water.
Notes
Hygroscopic. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 5 g |
| Beilstein | 3767479 |
| Sensitivity | Hygroscopic |
| Solubility Information | Partially soluble in cold water,hot water. |
| Formula Weight | 248.19 |
| Percent Purity | 99% |
| Assay | (dry wt.) |
| Chemical Name or Material | 1,3-Propanedisulfonic acid disodium salt |
Safety and Handling
| Hazard Category | H315-H318-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only