missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanedisulfonic acid disodium salt, 99% (dry wt.)

Catalog No. AAB2009206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2009206 5 g
AAB2009214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2009206 Supplier Thermo Scientific Chemicals Supplier No. B2009206
Only null left

CAS: 36589-58-9 | C3H6Na2O6S2 | 248.175 g/mol

It is used as a reactant involved in synthesis of cadmium(II) bipyridine complex metal-organic frameworks with alkanedisulfonates, alkylenedisulfonate inorganic-organic hybrid materials and glycosaminoglycan mimetic involved in pharmacological studies for the treatment of amyloid A amyloidosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reactant involved in synthesis of cadmium(II) bipyridine complex metal-organic frameworks with alkanedisulfonates, alkylenedisulfonate inorganic-organic hybrid materials and glycosaminoglycan mimetic involved in pharmacological studies for the treatment of amyloid A amyloidosis.

Solubility
Partially soluble in cold water, hot water.

Notes
Hygroscopic. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 36589-58-9
Molecular Formula C3H6Na2O6S2
Molecular Weight (g/mol) 248.175
MDL Number MFCD00191483
InChI Key DKGJFKPIUSHDIT-UHFFFAOYSA-L
Synonym eprodisate disodium, disodium 1,3-propanedisulfonate, kiacta, disodium propane-1,3-disulfonate, eprodisate sodium, unii-5x0d9h16iu, 1,3-propanedisulfonic acid disodium salt, eprodisate disodium usan, 1,3-propanedisulfonic acid, disodium salt
PubChem CID 6451143
IUPAC Name disodium;propane-1,3-disulfonate
SMILES C(CS(=O)(=O)[O-])CS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 3767479
Sensitivity Hygroscopic
Solubility Information Partially soluble in cold water,hot water.
Formula Weight 248.19
Percent Purity 99%
Assay (dry wt.)
Chemical Name or Material 1,3-Propanedisulfonic acid disodium salt
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only