missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanediol 98.0+%, TCI America™

Catalog No. p-7134631
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P048625G 25 g
P0486100G 100 g
P0486500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P048625G Supplier TCI America Supplier No. P048625G
Only null left

Chemical Identifiers

CAS 504-63-2
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
MDL Number MFCD00002949
InChI Key YPFDHNVEDLHUCE-UHFFFAOYSA-N
Synonym 1,3-propanediol, trimethylene glycol, 1,3-dihydroxypropane, 2-deoxyglycerol, 1,3-propylene glycol, 1,3-propylenediol, 1,3-propandiol, 2-hydroxymethyl ethanol, omega-propanediol, beta-propylene glycol
PubChem CID 10442
ChEBI CHEBI:16109
IUPAC Name propane-1,3-diol
SMILES OCCCO

Specifications

Color Colorless
Boiling Point 214°C
Quantity 25 g
Formula Weight 76.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Propanediol
EINECSNumber (2)-0234
RTECSNumber TY2010000
TSCA Yes