missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Propanediol di-p-toluenesulfonate, 98%, Thermo Scientific Chemicals
$98.49 - $98.49
Chemical Identifiers
| CAS | 5469-66-9 |
|---|---|
| Molecular Formula | C17H20O6S2 |
| Molecular Weight (g/mol) | 384.46 |
| MDL Number | MFCD00013392 |
| InChI Key | ODVREDGIWSDQFD-UHFFFAOYSA-N |
| Synonym | 1,3-bis tosyloxy propane, propane-1,3-diyl bis 4-methylbenzenesulfonate, 1,3-propanediol di-p-toluenesulfonate, 1,3-propanediol di-p-tosylate, 1,3-propanediol-di-4-tosylate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzene-1-sulfonate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzenesulfonate, propane-1,3-di-tos, acmc-209liv, 1,3-propanediol ditosylate |
| PubChem CID | 230370 |
| IUPAC Name | 3-(4-methylphenyl)sulfonyloxypropyl 4-methylbenzenesulfonate |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)OCCCOS(=O)(=O)C1=CC=C(C)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2108414
|
Thermo Scientific Chemicals
B2108414 |
25 g |
Each for $98.49
|
|
|||||
Description
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as pharmaceutical intermediate.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
| 5469-66-9 | |
| 384.46 | |
| ODVREDGIWSDQFD-UHFFFAOYSA-N | |
| 230370 | |
| CC1=CC=C(C=C1)S(=O)(=O)OCCCOS(=O)(=O)C1=CC=C(C)C=C1 |
| C17H20O6S2 | |
| MFCD00013392 | |
| 1,3-bis tosyloxy propane, propane-1,3-diyl bis 4-methylbenzenesulfonate, 1,3-propanediol di-p-toluenesulfonate, 1,3-propanediol di-p-tosylate, 1,3-propanediol-di-4-tosylate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzene-1-sulfonate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzenesulfonate, propane-1,3-di-tos, acmc-209liv, 1,3-propanediol ditosylate | |
| 3-(4-methylphenyl)sulfonyloxypropyl 4-methylbenzenesulfonate |
Specifications
| 5469-66-9 | |
| C17H20O6S2 | |
| 25 g | |
| ODVREDGIWSDQFD-UHFFFAOYSA-N | |
| 3-(4-methylphenyl)sulfonyloxypropyl 4-methylbenzenesulfonate | |
| 230370 | |
| 98% |
| 90°C to 92°C | |
| MFCD00013392 | |
| 1,3-bis tosyloxy propane, propane-1,3-diyl bis 4-methylbenzenesulfonate, 1,3-propanediol di-p-toluenesulfonate, 1,3-propanediol di-p-tosylate, 1,3-propanediol-di-4-tosylate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzene-1-sulfonate, 3-4-methylbenzenesulfonyl oxy propyl 4-methylbenzenesulfonate, propane-1,3-di-tos, acmc-209liv, 1,3-propanediol ditosylate | |
| CC1=CC=C(C=C1)S(=O)(=O)OCCCOS(=O)(=O)C1=CC=C(C)C=C1 | |
| 384.46 | |
| 384.47 | |
| 1,3-Propanediol di-p-toluenesulfonate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only