missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanediol, 98%

Catalog No. p-4509876
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131402500 250 mL Glass bottle
AC131400010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC131402500 Supplier Thermo Scientific Chemicals Supplier No. 131402500
Only null left

CAS: 504-63-2 | C3H8O2 | 76.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 504-63-2
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
MDL Number MFCD00002949
InChI Key YPFDHNVEDLHUCE-UHFFFAOYSA-N
Synonym 1,3-propanediol, trimethylene glycol, 1,3-dihydroxypropane, 2-deoxyglycerol, 1,3-propylene glycol, 1,3-propylenediol, 1,3-propandiol, 2-hydroxymethyl ethanol, omega-propanediol, beta-propylene glycol
PubChem CID 10442
ChEBI CHEBI:16109
IUPAC Name propane-1,3-diol
SMILES OCCCO

Specifications

Melting Point -32.0°C
Density 1.0520g/mL
Boiling Point 214.0°C
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4380 to 1.4400
Linear Formula HO(CH2)3OH
Quantity 250 mL
Beilstein 01,475
Fieser 13,268
Merck Index 15,9885
Specific Gravity 1.052
Solubility Information Solubility in water: 100g/L water. Other solubilities: soluble in ethanol, diethyl ether,, slightly soluble in benzene
Viscosity 52.7 mPa.s (20°C)
Formula Weight 76.09
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material 1, 3-Propanediol
Show More Show Less
EINECSNumber 207-997-3

RUO – Research Use Only