missing translation for 'onlineSavingsMsg'
Learn More

1,3-Phenylenediacetic Acid 98.0+%, TCI America™

Catalog No. P10645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10645G 5 g
P106425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10645G Supplier TCI America Supplier No. P10645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19806-17-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004341
InChI Key GDYYIJNDPMFMTB-UHFFFAOYSA-N
Synonym 2,2'-1,3-phenylene diacetic acid, 1,3-phenylenediacetic acid, m-benzenediacetic acid, m-phenylenediacetic acid, 1,3-benzenediacetic acid, acetic acid, 2,2'-m-phenylene di, 2-3-carboxymethyl phenyl acetic acid, 1,3-benzenediaceticacid, 3-carboxymethyl phenyl acetic acid, 1,3-benzoldiessigsaure
PubChem CID 29788
IUPAC Name 2-[3-(carboxymethyl)phenyl]acetic acid
SMILES OC(=O)CC1=CC(CC(O)=O)=CC=C1

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Phenylenediacetic Acid
RTECSNumber AJ2780000
TSCA No