missing translation for 'onlineSavingsMsg'
Learn More

1,3-Phenylenediacetic acid, 97%

Catalog No. p-7081445
Change view
Click to view available options
Quantity:
2 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1142904 2 g
AAA1142909 10 g
AAA1142914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1142904 Supplier Thermo Scientific Chemicals Supplier No. A1142904
Only null left

CAS: 19806-17-8 | C10H10O4 | 194.19 g/mol

In order to establish defined biomacromolecular systems for mineralised tissue applications, type I collagen was functionalised with 1,3-phenylenediacetic acid (Ph) and investigated at the molecular, macroscopic and functional levels. The solvothermal reactions of 1,2-bis(imidazol-1?-yl)ethane (bime), 1,3-phenylenediacetic acid, 1,4-phenylenediacetic acid), 1,2-phenylenediacetic acid with different metal ions gave rise to four novel helical coordination polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In order to establish defined biomacromolecular systems for mineralised tissue applications, type I collagen was functionalised with 1,3-phenylenediacetic acid (Ph) and investigated at the molecular, macroscopic and functional levels. The solvothermal reactions of 1,2-bis(imidazol-1′-yl)ethane (bime), 1,3-phenylenediacetic acid, 1,4-phenylenediacetic acid), 1,2-phenylenediacetic acid with different metal ions gave rise to four novel helical coordination polymers.

Solubility
Slightly soluble in water. Solublitiy in methanol is almost transparency.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 19806-17-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004341
InChI Key GDYYIJNDPMFMTB-UHFFFAOYSA-N
Synonym 2,2'-1,3-phenylene diacetic acid, 1,3-phenylenediacetic acid, m-benzenediacetic acid, m-phenylenediacetic acid, 1,3-benzenediacetic acid, acetic acid, 2,2'-m-phenylene di, 2-3-carboxymethyl phenyl acetic acid, 1,3-benzenediaceticacid, 3-carboxymethyl phenyl acetic acid, 1,3-benzoldiessigsaure
PubChem CID 29788
IUPAC Name 2-[3-(carboxymethyl)phenyl]acetic acid
SMILES OC(=O)CC1=CC(CC(O)=O)=CC=C1

Specifications

Melting Point 172°C to 178°C
Odor Odorless
Quantity 2 g
Beilstein 2616749
Solubility Information Slightly soluble in water. Solublitiy in methanol is almost transparency.
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 1,3-Phenylenediacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-332-3
RTECSNumber AJ2780000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only