missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Methoxyphenyl)piperazine 98.0+%, TCI America™

Catalog No. M21355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21355G 5 g
M213525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21355G Supplier TCI America Supplier No. M21355G

Chemical Identifiers

CAS 16015-71-7
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD00040733
InChI Key PZIBVWUXWNYTNL-UHFFFAOYSA-N
Synonym 1-3-methoxyphenyl piperazine, n-3-methoxyphenyl piperazine, 1-3-methoxyphenyl-piperazine, unii-6lcp52962g, 1-3-methoxy-phenyl-piperazine, chembl59597, piperazine, 1-3-methoxyphenyl, 3-methoxy-1-piperazinylbenzene, pubchem8585, 3-meopp
PubChem CID 81430
IUPAC Name 1-(3-methoxyphenyl)piperazine
SMILES COC1=CC=CC(=C1)N2CCNCC2

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 g
UN Number 3267
Formula Weight 192.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(3-Methoxyphenyl)piperazine