missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Methoxyphenyl)piperazine dihydrochloride, 98%

Catalog No. AC126050250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126050250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC126050250 Supplier Thermo Scientific Chemicals Supplier No. 126050250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6968-76-9 | C11H16N22ClH | 265.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6968-76-9
Molecular Formula C11H16N22ClH
Molecular Weight (g/mol) 265.18
MDL Number MFCD00035285
InChI Key UKUNKQNESKRETR-UHFFFAOYSA-N
Synonym 1-3-methoxyphenyl piperazine dihydrochloride, unii-q0t82qhg7n, 1-3-methoxyphenyl piperazine 2hcl, q0t82qhg7n, piperazine, 1-3-methoxyphenyl-, dihydrochloride, 1-3-methoxyphenyl piperazindihydrochlorid, 1-3-methoxyphenyl piperazine hcl, pubchem15280, 1-3-methoxyphenyl piperazinehydrochloride, n-3-methoxyphenyl piperazine dihydrochloride
PubChem CID 81429
IUPAC Name 1-(3-methoxyphenyl)piperazine;dihydrochloride
SMILES COC1=CC=CC(=C1)N2CCNCC2.Cl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 201°C to 204°C
Color Gray to White or Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 265.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Methoxyphenyl)piperazine dihydrochloride
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-189-7

RUO – Research Use Only