missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Indanedione, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.
Solubility
Soluble in ethanol, ether, benzene, slightly soluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
It is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.
Solubility
Soluble in ethanol, ether, benzene, slightly soluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Specifications
Specifications
| CAS | 606-23-5 |
| Density | 1.37 |
| Molecular Formula | C9H6O2 |
| Quantity | 100g |
| MDL Number | MFCD00003779 |
| Beilstein | 1210061 |
| Synonym | 1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g |
| Solubility Information | Soluble in ethanol,ether,benzene,slightly soluble in water. |
| InChI Key | UHKAJLSKXBADFT-UHFFFAOYSA-N |
| SMILES | C1C(=O)C2=CC=CC=C2C1=O |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 210-109-7 |
| RTECSNumber | NK5070000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only