missing translation for 'onlineSavingsMsg'
Learn More

1,3-Indanedione, 97%

Catalog No. AAA1220614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1220614 25 g
AAA1220606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1220614 Supplier Thermo Scientific Chemicals Supplier No. A1220614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 606-23-5 | C9H6O2 | 146.145 g/mol

It is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.

Solubility
Soluble in ethanol, ether, benzene, slightly soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 606-23-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.145
MDL Number MFCD00003779
InChI Key UHKAJLSKXBADFT-UHFFFAOYSA-N
Synonym 1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g
PubChem CID 11815
ChEBI CHEBI:78877
IUPAC Name indene-1,3-dione
SMILES C1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 128°C to 132°C
Density 1.37
Quantity 25 g
Beilstein 1210061
Solubility Information Soluble in ethanol,ether,benzene,slightly soluble in water.
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material 1,3-Indanedione
Hazard Category H302-H315-H319
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-109-7
RTECSNumber NK5070000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only