missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Hydroxypropyl)piperazine, 98%

Catalog No. AAH5029003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5029003 1 g
AAH5029006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5029003 Supplier Thermo Scientific Chemicals Supplier No. H5029003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5317-32-8 | C7H16N2O | 144.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5317-32-8
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.22
MDL Number MFCD00023132
InChI Key LWEOFVINMVZGAS-UHFFFAOYSA-N
Synonym 3-piperazin-1-yl propan-1-ol, 1-piperazinepropanol, 1-3-hydroxypropyl piperazine, 1-3-hydroxypropyl-piperazine, piperazin-1-ylpropanol, 3-1-piperazinyl-1-propanol, 1-3-hydroxypropyl-4-piperazine, hydroxypropyl-piperazin, acmc-20a7ij, piperazin-1-ylpropan-ol
PubChem CID 79207
IUPAC Name 3-piperazin-1-ylpropan-1-ol
SMILES OCCCN1CCNCC1

Specifications

Melting Point 48°C to 52°C
Quantity 1 g
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material 1-(3-Hydroxypropyl)piperazine
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 226-176-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only