missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3-Hydroxypropyl)-2-pyrrolidone 96.0+%, TCI America™
$106.25 - $216.24
Chemical Identifiers
| CAS | 62012-15-1 |
|---|---|
| Molecular Formula | C7H13NO2 |
| Molecular Weight (g/mol) | 143.19 |
| MDL Number | MFCD00059014 |
| InChI Key | CVDGNRZPDAXOQO-UHFFFAOYSA-N |
| PubChem CID | 566744 |
| IUPAC Name | 1-(3-hydroxypropyl)pyrrolidin-2-one |
| SMILES | OCCCN1CCCC1=O |
Chemical Identifiers
| 62012-15-1 | |
| 143.19 | |
| CVDGNRZPDAXOQO-UHFFFAOYSA-N | |
| 1-(3-hydroxypropyl)pyrrolidin-2-one |
| C7H13NO2 | |
| MFCD00059014 | |
| 566744 | |
| OCCCN1CCCC1=O |
Specifications
| 62012-15-1 | |
| 140°C | |
| MFCD00059014 | |
| CVDGNRZPDAXOQO-UHFFFAOYSA-N | |
| 1-(3-hydroxypropyl)pyrrolidin-2-one | |
| 566744 | |
| ≥96.0% (GC) | |
| 1-(3-Hydroxypropyl)-2-pyrrolidone |
| Yellow | |
| C7H13NO2 | |
| 10 mL | |
| OCCCN1CCCC1=O | |
| 143.19 | |
| 143.19 | |
| Liquid |
Safety and Handling
TSCA : No