missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Hydroxypropyl)-2-pyrrolidone 96.0+%, TCI America™

Catalog No. H054110ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H054110ML 10 mL
H054125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H054110ML Supplier TCI America Supplier No. H054110ML

Chemical Identifiers

CAS 62012-15-1
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00059014
InChI Key CVDGNRZPDAXOQO-UHFFFAOYSA-N
PubChem CID 566744
IUPAC Name 1-(3-hydroxypropyl)pyrrolidin-2-one
SMILES OCCCN1CCCC1=O

Specifications

Color Yellow
Boiling Point 140°C
Quantity 10 mL
Formula Weight 143.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(3-Hydroxypropyl)-2-pyrrolidone