missing translation for 'onlineSavingsMsg'
Learn More

1,3-Divinyltetramethyldisiloxane 98.0+%, TCI America™

Catalog No. p-7126923
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D178025ML 25 mL
D1780100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D178025ML Supplier TCI America Supplier No. D178025ML

Chemical Identifiers

CAS 2627-95-4
Molecular Formula C8H18OSi2
Molecular Weight (g/mol) 186.40
MDL Number MFCD00014933
InChI Key BITPLIXHRASDQB-UHFFFAOYSA-N
Synonym 1,3-divinyltetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, disiloxane, 1,3-diethenyl-1,1,3,3-tetramethyl, 1,1'-divinyltetramethyldisiloxane, unii-sj4d71zqf4, bisvinyltetramethyldisiloxane, 1,3-divinyl-1,1,3,3-tetramethyl disiloxane, sym-tetramethyldivinyldisiloxane, tetramethyl-1,3-divinyldisiloxane, 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane
PubChem CID 75819
IUPAC Name ethenyl[(ethenyldimethylsilyl)oxy]dimethylsilane
SMILES C[Si](C)(O[Si](C)(C)C=C)C=C

Specifications

Color Colorless
Boiling Point 139°C
Quantity 25 mL
UN Number 1993
Formula Weight 186.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Divinyltetramethyldisiloxane
EINECSNumber (7)-0483
RTECSNumber JM9235250
TSCA Yes